accessibility menu, dialog, popup

UserName

H3CCH3ONHOHONCH3H3C(E)(E)

CAS RN 58880-19-6

IUPAC Name: (2E,4E)-7-[4-(dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxohepta-2,4-dienamide
Synonyms: trichostatin A
Molecular Weight (g/mol): 302.37
Molecular Formula: C17H22N2O3
InChi Key: RTKIYFITIVXBLE-WKWSCTOINA-N
SMILES: CC(\C=C(/C)\C=C\C(=O)NO)C(=O)C1=CC=C(C=C1)N(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results