accessibility menu, dialog, popup

UserName

OHN(E/Z)

CAS RN 588-53-4

IUPAC Name: 4-[(phenylmethylidene)amino]phenol
Synonyms: phenol, p-(benzylideneamino)-
Molecular Weight (g/mol): 197.24
Molecular Formula: C13H11NO
InChi Key: BVTLIIQDQAUXOI-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)N=CC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results