accessibility menu, dialog, popup

UserName

CH3CH3H3CH3CH3CCH3

CAS RN 58775-05-6

IUPAC Name: 2,7-di-tert-butyl-9H-fluorene
Synonyms: 2,7-di-tert-butyl-9H-fluorene
Molecular Weight (g/mol): 278.44
Molecular Formula: C21H26
InChi Key: DFZYPLLGAQIQTD-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC2=C(C=C1)C1=C(C2)C=C(C=C1)C(C)(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)