accessibility menu, dialog, popup

UserName

CAS RN 585-76-2

IUPAC Name: 3-bromobenzoic acid
Synonyms: M-bromobenzoic acid
Molecular Weight (g/mol): 201.02
Molecular Formula: C7H5BrO2
InChi Key: VOIZNVUXCQLQHS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(Br)=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)