missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzoic acid, 98+%

Catalog No. AAA1260622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1260622 100 g
AAA1260614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1260622 Supplier Thermo Scientific Chemicals Supplier No. A1260622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 585-76-2 | C7H5BrO2 | 201.019 g/mol

3-Bromobenzoic acid was used as test solute in the determination of acidity constants by capillary zone electrophoresis. It was used as internal standard to study the retention mechanisms of an unmodified and a hydroxylated polystyrene-divinylbenzene polymer by solid-phase extraction. It was also used in synthesis of N-(1,1-dimethyl-2-hydroxyethyl)-3-bromobenzamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobenzoic acid was used as test solute in the determination of acidity constants by capillary zone electrophoresis. It was used as internal standard to study the retention mechanisms of an unmodified and a hydroxylated polystyrene-divinylbenzene polymer by solid-phase extraction. It was also used in synthesis of N-(1,1-dimethyl-2-hydroxyethyl)-3-bromobenzamide.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 585-76-2
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.019
MDL Number MFCD00002487
InChI Key VOIZNVUXCQLQHS-UHFFFAOYSA-N
Synonym m-bromobenzoic acid, 3-brombenzoic acid, benzoic acid, 3-bromo, 3-bromo-benzoic acid, benzoic acid, m-bromo, unii-7dct3a73lx, 7dct3a73lx, 3-bromo benzene acid, 3-bromobenzoicacid, 4-fluorobenzoicacid
PubChem CID 11456
IUPAC Name 3-bromobenzoic acid
SMILES C1=CC(=CC(=C1)Br)C(=O)O

Specifications

Melting Point 153°C to 157°C
Density 1.948
Quantity 100 g
Beilstein 2041792
Solubility Information Insoluble in water.
Formula Weight 201.02
Percent Purity ≥98%
Chemical Name or Material 3-Bromobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-562-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only