missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzoic Acid 98.0+%, TCI America™

Catalog No. B055125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B055125G 25 g
B0551250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B055125G Supplier TCI America Supplier No. B055125G

Chemical Identifiers

CAS 585-76-2
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.019
MDL Number MFCD00002487
InChI Key VOIZNVUXCQLQHS-UHFFFAOYSA-N
Synonym m-bromobenzoic acid, 3-brombenzoic acid, benzoic acid, 3-bromo, 3-bromo-benzoic acid, benzoic acid, m-bromo, unii-7dct3a73lx, 7dct3a73lx, 3-bromo benzene acid, 3-bromobenzoicacid, 4-fluorobenzoicacid
PubChem CID 11456
IUPAC Name 3-bromobenzoic acid
SMILES C1=CC(=CC(=C1)Br)C(=O)O

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 25 g
Formula Weight 201.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromobenzoic Acid
EINECSNumber (3)-0062
TSCA Yes