accessibility menu, dialog, popup

UserName

CAS RN 5826-73-3

IUPAC Name: 2,3-dimethyl bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate
Synonyms: dimalone
Molecular Weight (g/mol): 210.23
Molecular Formula: C11H14O4
InChi Key: VGQLNJWOULYVFV-UHFFFAOYNA-N
SMILES: COC(=O)C1C2CC(C=C2)C1C(=O)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results