CAS RN 582-65-0
IUPAC Name:
4,4,4-trifluoro-1-(4-fluorophenyl)butane-1,3-dione
Synonyms:
4,4,4-trifluoro-1-(4-fluorophenyl)butane-1,3-dione
Molecular Weight (g/mol):
234.15
Molecular Formula:
C10H6F4O2
InChi Key:
KEZLARPKXOHKJS-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C=C1)C(=O)CC(=O)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
3-(4-Fluorobenzoyl)-1,1,1-trifluoroacetone, 97%
3-(4-Fluorobenzoyl)-1,1,1-trifluoroacetone, 97%
CAS: 582-65-0 Molecular Formula: C10H6F4O2 Molecular Weight (g/mol): 234.15 MDL Number: MFCD03419766 InChI Key: KEZLARPKXOHKJS-UHFFFAOYSA-N Synonym: 4,4,4-trifluoro-1-4-fluorophenyl butane-1,3-dione,4,4,4-trifluoro-1-4-fluorophenyl-1,3-butanedione,3-4-fluorobenzoyl-1,1,1-trifluoroacetone,1,3-butanedione, 4,4,4-trifluoro-1-4-fluorophenyl,4,4,4-trifluoro-1-4-fluoro-phenyl-butane-1,3-dione,1-4'-fluorophenyl-4,4,4-trifluoro-1,3-butanedione,tos-bb-0670,1-4-fluorophenyl-4,4,4-trifluoro-1,3-butanedione,1-4-fluoro-phenyl-4,4,4-trifluoro-butane-1,3-dione PubChem CID: 2771476 IUPAC Name: 4,4,4-trifluoro-1-(4-fluorophenyl)butane-1,3-dione SMILES: FC1=CC=C(C=C1)C(=O)CC(=O)C(F)(F)F