accessibility menu, dialog, popup

UserName

ClCH3NH3CH3CH3C

CAS RN 5810-42-4

IUPAC Name: tetrapropylazanium chloride
Synonyms: tetrapropylammonium chloride
Molecular Weight (g/mol): 221.81
Molecular Formula: C12H28ClN
InChi Key: FBEVECUEMUUFKM-UHFFFAOYSA-M
SMILES: [Cl-].CCC[N+](CCC)(CCC)CCC
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)