accessibility menu, dialog, popup

UserName

H2NSOOHNNHSOOCl

CAS RN 58-93-5

IUPAC Name: 6-chloro-1,1-dioxo-3,4-dihydro-2H-1λ⁶,2,4-benzothiadiazine-7-sulfonamide
Synonyms: hydrochlorothiazide dichlorotride capozide dihydrochlorothiazid neo-codema dichlotiazid ro-hydrazide bremil hidrotiazida roxane
Molecular Weight (g/mol): 297.73
Molecular Formula: C7H8ClN3O4S2
InChi Key: JZUFKLXOESDKRF-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC2=C(NCNS2(=O)=O)C=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results