accessibility menu, dialog, popup

UserName

CAS RN 58-58-2

IUPAC Name: (2S)-2-amino-N-[(2S,3S,4R,5R)-5-[6-(dimethylamino)-9H-purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide dihydrochloride
Synonyms: puromycin hydrochloride
Molecular Weight (g/mol): 544.43
Molecular Formula: C22H31Cl2N7O5
InChi Key: MKSVFGKWZLUTTO-MLYJQVMKNA-N
SMILES: Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1
Physical Form 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Type 
  • (6)
Color 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
  • (3)
  • (1)
  • (3)