missing translation for 'onlineSavingsMsg'
Learn More

Puromycin Dihydrochloride, Fisher BioReagents™

Catalog No. BP2956100
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP2956100 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP2956100 Supplier Fisher BioReagents Supplier No. BP2956 100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibits protein synthesis | CAS: 58-58-2 | C22H31Cl2N7O5 | 544.43 g/mol

Inhibits protein synthesis

Specifications

Type Puromycin Dihydrochloride
Chemical Name or Material Puromycin Dihydrochloride
Physical Form Solid
CAS 58-58-2
Melting Point 168°C
Source Streptomyces alboniger
Packaging Clear Glass
Water <12.0
Color Beige
Quantity 100 mg
Assay Percent Range > 98 %
Molecular Formula C22H31Cl2N7O5
MDL Number MFCD00150080
Merck Index 15, 8054
InChI Key MKSVFGKWZLUTTO-MLYJQVMKNA-N
SMILES Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1
IUPAC Name (2S)-2-amino-N-[(2S,3S,4S,5R)-5-[6-(dimethylamino)purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide;dihydrochloride
Molecular Weight (g/mol) 544.43
PubChem CID 131632508
Formula Weight 544.44
Percent Purity 98% min.
Show More Show Less