accessibility menu, dialog, popup

UserName

H3CONCH3H3COOH3C

CAS RN 58-46-8

IUPAC Name: 9,10-dimethoxy-3-(2-methylpropyl)-1H,2H,3H,4H,6H,7H,11bH-pyrido[2,1-a]isoquinolin-2-one
Synonyms: tetrabenazine roche brand of tetrabenazine orphan brand of tetrabenazine tetrabenazine product (product) tetrabenazine orphan brand nitoman shire brand of tetrabenazine tetrabenazine (inn) and its salts tetrabenazine product cambridge laboratories brand of tetrabenazine
Molecular Weight (g/mol): 317.43
Molecular Formula: C19H27NO3
InChi Key: MKJIEFSOBYUXJB-UHFFFAOYNA-N
SMILES: COC1=CC2=C(C=C1OC)C1CC(=O)C(CC(C)C)CN1CC2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results