missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabenazine, MilliporeSigma™ Supelco™
Description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.
Specifications
Specifications
| CAS | 58-46-8 |
| Purity Grade Notes | These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind. |
| Type | USP Reference Standard |
| Quantity | 200 mg |
| Grade | Pharmaceutical Primary Standard |