missing translation for 'onlineSavingsMsg'
Learn More

Tetrabenazine 99.0+%, TCI America™

Catalog No. T2839200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2839200MG 200 mg
T28391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2839200MG Supplier TCI America Supplier No. T2839200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58-46-8
Molecular Formula C19H27NO3
Molecular Weight (g/mol) 317.429
InChI Key MKJIEFSOBYUXJB-UHFFFAOYSA-N
Synonym 3-Isobutyl-9,10-dimethoxy-1,3,4,6,7,11b-hexahydro-2H-benzo[a]quinolizin-2-one
PubChem CID 6018
ChEBI CHEBI:64028
IUPAC Name 9,10-dimethoxy-3-(2-methylpropyl)-1,3,4,6,7,11b-hexahydrobenzo[a]quinolizin-2-one
SMILES CC(C)CC1CN2CCC3=CC(=C(C=C3C2CC1=O)OC)OC

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 200 mg
Formula Weight 317.43
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabenazine
EINECSNumber (9)-2283
RTECSNumber DK2275000
TSCA No
Recommended Storage Refrigerator