accessibility menu, dialog, popup

UserName

CAS RN 58-14-0

IUPAC Name: 5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine
Synonyms: pyrimethamine maloprim pyrimethaminum malacid diaminopyritamin daraprime darachlor pyrimethamin daraprim darapram
Molecular Weight (g/mol): 248.71
Molecular Formula: C12H13ClN4
InChi Key: WKSAUQYGYAYLPV-UHFFFAOYSA-N
SMILES: CCC1=NC(N)=NC(N)=C1C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)