accessibility menu, dialog, popup

UserName

CAS RN 5791-00-4

IUPAC Name: 6-chloro-2-methyl-3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms: 6-chloro-2-methyl-2,4-dihydro-1,4-benzoxazin-3-one
Molecular Weight (g/mol): 197.62
Molecular Formula: C9H8ClNO2
InChi Key: MGOCBXKZDZRPMK-UHFFFAOYNA-N
SMILES: CC1OC2=CC=C(Cl)C=C2NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results