accessibility menu, dialog, popup

UserName

CH3CH3H2N

CAS RN 579-66-8

IUPAC Name: 2,6-diethylaniline
Synonyms: 2,6-diethylaniline DEA
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: FOYHNROGBXVLLX-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(CC)=C1N
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)