accessibility menu, dialog, popup

UserName

CAS RN 57709-61-2

IUPAC Name: 1,10-phenanthroline-2,9-dicarboxylic acid
Synonyms: 1,10-phenanthroline-2,9-dicarboxylic acid
Molecular Weight (g/mol): 268.23
Molecular Formula: C14H8N2O4
InChi Key: FXSVCROWUPWXBP-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC2=C(C=CC3=C2N=C(C=C3)C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results