accessibility menu, dialog, popup

UserName

ONOBr

CAS RN 577-19-5

IUPAC Name: 1-bromo-2-nitrobenzene
Synonyms: O-bromonitrobenzene
Molecular Weight (g/mol): 202.01
Molecular Formula: C6H4BrNO2
InChi Key: ORPVVAKYSXQCJI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1Br
Molecular Weight (g/mol) 
  • (7)
  • (1)
Boiling Point 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (4)
  • (7)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)