accessibility menu, dialog, popup

UserName

CAS RN 57508-48-2

IUPAC Name: (1-amino-3-ethoxy-3-oxopropylidene)azanium
Synonyms: (1-amino-3-ethoxy-3-oxopropylidene)azanium
Molecular Weight (g/mol): 131.15
Molecular Formula: C5H11N2O2
InChi Key: HSDKTLKBDJXJQU-UHFFFAOYSA-O
SMILES: CCOC(=O)CC(N)=[NH2+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results