accessibility menu, dialog, popup

UserName

CAS RN 575-38-2

IUPAC Name: naphthalene-1,7-diol
Synonyms: napthalene-1,7-diol
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: ZUVBIBLYOCVYJU-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=CC=C(O)C2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)