CAS RN 5736-91-4
IUPAC Name:
4-(pentyloxy)benzaldehyde
Synonyms:
4-(pentyloxy)benzaldehyde
Molecular Weight (g/mol):
192.26
Molecular Formula:
C12H16O2
InChi Key:
YAPVGSXODFOBBR-UHFFFAOYSA-N
SMILES:
CCCCCOC1=CC=C(C=O)C=C1
1
–
1
of
1
results
1
4-Amyloxybenzaldehyde 98.0+%, TCI America™
CAS: 5736-91-4 Molecular Formula: C12H16O2 Molecular Weight (g/mol): 192.258 MDL Number: MFCD00014135 InChI Key: YAPVGSXODFOBBR-UHFFFAOYSA-N Synonym: 4-pentyloxy benzaldehyde,4-amyloxybenzaldehyde,p-pentyloxybenzaldehyde,benzaldehyde, 4-pentyloxy,4-pentyloxybenzaldehyde,benzaldehyde, p-pentyloxy,p-amyloxy benzaldehyde,4-n-amyloxybenzaldehyde,4-n-pentyloxybenzaldehyde,p-pentyloxy benzaldehyde PubChem CID: 79815 IUPAC Name: 4-pentoxybenzaldehyde SMILES: CCCCCOC1=CC=C(C=C1)C=O