accessibility menu, dialog, popup

UserName

CAS RN 5735-53-5

IUPAC Name: 3,4-dihydro-2H-1,4-benzoxazine
Synonyms: 3,4-dihydro-2H-1,4-benzoxazine
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: YRLORWPBJZEGBX-UHFFFAOYSA-N
SMILES: C1COC2=CC=CC=C2N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results