accessibility menu, dialog, popup

UserName

BrBrBrBr

CAS RN 570414-33-4

IUPAC Name: 2,7-dibromo-9,9-bis(6-bromohexyl)-9H-fluorene
Synonyms: 2,7-dibromo-9,9-bis(6-bromohexyl)fluorene
Molecular Weight (g/mol): 650.13
Molecular Formula: C25H30Br4
InChi Key: OJMAUBALNSWGDC-UHFFFAOYSA-N
SMILES: BrCCCCCCC1(CCCCCCBr)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results