accessibility menu, dialog, popup

UserName

BrH3CNCH3CH3H3C

CAS RN 57-09-0

IUPAC Name: hexadecyltrimethylazanium bromide
Synonyms: cetyltrimethylammonium bromide|cetrimide|cee dee|cetrimonium bromide|ctmab|cetab|centimide|bromure de cetrimonium|bromuro de cetrimonio|cetrimonii bromidum
Molecular Weight (g/mol): 364.46
Molecular Formula: C19H42BrN
InChi Key: LZZYPRNAOMGNLH-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (24)
Quantity 
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (6)
  • (2)
  • (2)
  • (3)
  • (4)
Physical Form 
  • (3)
  • (2)
  • (2)
  • (4)
Grade 
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (4)
  • (1)
  • (2)
  • (1)
  • (6)
  • (4)
  • (3)
  • (9)
  • (2)
  • (24)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)