missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Hexadecyltrimethylammonium bromide, ≥99.0% (AT), MilliporeSigma™ Supelco™

Catalog No. 111018728
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018728 10 g
111018023 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111018728 Supplier Merck Emd Millipore Supplier No. 5236710GF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hexadecyltrimethylammonium bromide (CTAB) is a quaternary ammonium salt containing one long hydrophobic alkyl chain called amphiphile. The hydrophobic ions interact forming discrete aggregates possessing a hydrophobic core and a polar surface, called micelles in polar solution.

Chemical Identifiers

MDL Number MFCD00011772
Synonym CTAB; Cetrimonium bromide; Cetyltrimethylammonium bromide; Palmityltrimethylammonium bromide

Specifications

Melting Point 248°C to 251°C
CAS 57-09-0
Physical Form Powder or Crystals
Quantity 10 g
Linear Formula CH3(CH2)15 N(Br)(CH3)3
Flash Point 244°C
Solubility Information Water: soluble 36.4 g/L (at 20°C (completely))
Molecular Weight (g/mol) 364.45
Formula Weight 364.45
Percent Purity ≥99.0% (AT)
Grade Ion Pair Chromatography
Packaging Bottle
Product Line LiChropur
Chemical Name or Material Hexadecyltrimethylammonium Bromide
Show More Show Less
RTECSNumber BQ7875000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany