accessibility menu, dialog, popup

UserName

ClClClO

CAS RN 56961-75-2

IUPAC Name: 2,3,5-trichlorobenzaldehyde
Synonyms: 2,3,5-trichlorobenzaldehyde
Molecular Weight (g/mol): 209.45
Molecular Formula: C7H3Cl3O
InChi Key: DJYRZTCLVDKWBL-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(Cl)C(C=O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results