missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trichlorobenzaldehyde 95.0+%, TCI America™

Catalog No. T11825G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T11825G Supplier TCI America Supplier No. T11825G
Only null left

Chemical Identifiers

CAS 56961-75-2
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00010860
InChI Key DJYRZTCLVDKWBL-UHFFFAOYSA-N
Synonym benzaldehyde, 2,3,5-trichloro, ksc606a4b, acmc-1b126, benzaldehyde,2,3,5-trichloro, djyrztclvdkwbl-uhfffaoysa, 2,3,5-trichlorobenzaldehyde
PubChem CID 92596
IUPAC Name 2,3,5-trichlorobenzaldehyde
SMILES ClC1=CC(Cl)=C(Cl)C(C=O)=C1

Specifications

Melting Point 75°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 209.45
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5-Trichlorobenzaldehyde