accessibility menu, dialog, popup

UserName

OHOHOCl

CAS RN 56961-30-9

IUPAC Name: 2-chloro-5-hydroxybenzoic acid
Synonyms: 2-chloro-5-hydroxybenzoic acid
Molecular Weight (g/mol): 172.56
Molecular Formula: C7H5ClO3
InChi Key: UTVCLUZQPSRKMY-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=CC(O)=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)