missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-hydroxybenzoic acid, 99%, Thermo Scientific Chemicals

Catalog No. AAH2611203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2611203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2611203 Supplier Thermo Scientific Chemicals Supplier No. H2611203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56961-30-9 | Molecular Formula: C7H5ClO3 | Molecular Weight: 172.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56961-30-9
Molecular Formula C7H5ClO3
Molecular Weight (g/mol) 172.56
MDL Number MFCD04038818
InChI Key UTVCLUZQPSRKMY-UHFFFAOYSA-N
Synonym 2-chloro-5-hydroxybenzoicacid, benzoic acid, 2-chloro-5-hydroxy, 3-carboxy-4-chlorophenol, 2-chloro-5-hydroxybenzenecarboxylic acid, 2-chloro-5-hydroxy-benzoic acid, 2-chloro-5-hydroxy benzoic acid, 2-chloro-5-hydroxybenzene carboxylic acid, pubchem16487, acmc-1au79, ksc269a4t
PubChem CID 458210
IUPAC Name 2-chloro-5-hydroxybenzoic acid
SMILES OC(=O)C1=C(Cl)C=CC(O)=C1

Specifications

Melting Point 178°C to 180°C
Quantity 1 g
Formula Weight 172.57
Percent Purity 99%
Chemical Name or Material 2-Chloro-5-hydroxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only