accessibility menu, dialog, popup

UserName

HClCH3OOH2N

CAS RN 5680-79-5

IUPAC Name: hydrogen methyl 2-aminoacetate chloride
Synonyms: hydrogen methyl 2-aminoacetate chloride
Molecular Weight (g/mol): 125.55
Molecular Formula: C3H8ClNO2
InChi Key: COQRGFWWJBEXRC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC(=O)CN
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)