accessibility menu, dialog, popup

UserName

CAS RN 56741-94-7

NH2NOHN

CAS RN 56741-94-7

IUPAC Name: 2-amino-6-phenyl-1,4-dihydropyrimidin-4-one
Synonyms: 2-amino-6-phenyl-1H-pyrimidin-4-one
Molecular Weight (g/mol): 187.2
Molecular Formula: C10H9N3O
InChi Key: KXYGHDWFGFZJPJ-UHFFFAOYSA-N
SMILES: NC1=NC(=O)C=C(N1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results