accessibility menu, dialog, popup

UserName

CAS RN 56518-42-4

IUPAC Name: 4-bromo-3,5-dimethoxybenzoate
Synonyms: 4-bromo-3,5-dimethoxybenzoate
Molecular Weight (g/mol): 260.06
Molecular Formula: C9H8BrO4
InChi Key: JNFZULSIYYVRJO-UHFFFAOYSA-M
SMILES: COC1=CC(=CC(OC)=C1Br)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)