accessibility menu, dialog, popup

UserName

CAS RN 5616-32-0

IUPAC Name: 2-(methylamino)acetonitrile
Synonyms: 2-(methylamino)acetonitrile
Molecular Weight (g/mol): 70.1
Molecular Formula: C3H6N2
InChi Key: PVVRRUUMHFWFQV-UHFFFAOYSA-N
SMILES: CNCC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results