accessibility menu, dialog, popup

UserName

CAS RN 56057-19-3

IUPAC Name: 2-chloro-6-methyl-3-nitropyridine
Synonyms: 2-chloro-6-methyl-3-nitropyridine
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: UIEVSGOVFXWCIK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(Cl)=N1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)