accessibility menu, dialog, popup

UserName

CH3OONOH

CAS RN 56026-36-9

IUPAC Name: methyl 6-(hydroxymethyl)pyridine-3-carboxylate
Synonyms: methyl 6-(hydroxymethyl)pyridine-3-carboxylate
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: QVLONHSXRWCCAE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=C(CO)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results