accessibility menu, dialog, popup

UserName

CAS RN 56-55-3

IUPAC Name: tetraphene
Synonyms: benz[a]anthracene naphthanthracene 1,2-benz(a)anthracene benzanthracene benz(a)anthracene benzo(a)anthracene benzoanthracene 1,2-benzoanthracene 2,3-benzphenanthrene benzo(b)phenanthrene
Molecular Weight (g/mol): 228.29
Molecular Formula: C18H12
InChi Key: DXBHBZVCASKNBY-UHFFFAOYSA-N
SMILES: C1=CC=C2C=C3C(C=CC4=CC=CC=C34)=CC2=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)