accessibility menu, dialog, popup

UserName

HHBrBrH2NSNH2NH

CAS RN 56-10-0

IUPAC Name: dihydrogen [(2-aminoethyl)sulfanyl]methanimidamide dibromide
Synonyms: dihydrogen (2-aminoethyl)sulfanylmethanimidamide dibromide
Molecular Weight (g/mol): 281.01
Molecular Formula: C3H11Br2N3S
InChi Key: XDVMCVGTDUKDHL-UHFFFAOYSA-N
SMILES: [H+].[H+].[Br-].[Br-].NCCSC(N)=N
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)