missing translation for 'onlineSavingsMsg'
Learn More

S-(2-Aminoethyl)isothiourea dihydrobromide, 98%

Catalog No. AAL1423214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1423214 25 g
AAL1423206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1423214 Supplier Thermo Scientific Chemicals Supplier No. L1423214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56-10-0 | C3H11Br2N3S | 281.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-10-0
Molecular Formula C3H11Br2N3S
Molecular Weight (g/mol) 281.01
MDL Number MFCD00037011
InChI Key XDVMCVGTDUKDHL-UHFFFAOYSA-N
Synonym 2-2-aminoethyl isothiourea dihydrobromide, antiradon, antirad, surrectan, ixecur, 2-aminoethyl carbamimidothioate dihydrobromide, usaf xr-31, 2-2-aminoethyl-2-thiopseudourea dihydrobromide, 2-aminoethylisothiouronium dibromide, carbamimidothioic acid, 2-aminoethyl ester, dihydrobromide
PubChem CID 5940
IUPAC Name 2-aminoethyl carbamimidothioate;dihydrobromide
SMILES C(CSC(=N)N)N.Br.Br

Specifications

Melting Point 190°C to 192°C
Odor Odorless
Quantity 25 g
Beilstein 3911163
Merck Index 14,178
Formula Weight 281.02
Percent Purity 98%
Chemical Name or Material S-(2-Aminoethyl)isothiourea dihydrobromide
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P280f
Wear protective clothing.
EINECSNumber 200-257-0
RTECSNumber UM0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only