CAS RN 5599-50-8
IUPAC Name:
3,6-dimethyl-9H-carbazole
Synonyms:
3,6-dimethyl-9H-carbazole
Molecular Weight (g/mol):
195.27
Molecular Formula:
C14H13N
InChi Key:
HNACKJNPFWWEKI-UHFFFAOYSA-N
SMILES:
CC1=CC2=C(NC3=C2C=C(C)C=C3)C=C1
1
–
1
of
1
results
1
3,6-Dimethylcarbazole 98.0+%, TCI America™
CAS: 5599-50-8 Molecular Formula: C14H13N Molecular Weight (g/mol): 195.27 MDL Number: MFCD00814049 InChI Key: HNACKJNPFWWEKI-UHFFFAOYSA-N PubChem CID: 606965 IUPAC Name: 3,6-dimethyl-9H-carbazole SMILES: CC1=CC2=C(NC3=C2C=C(C)C=C3)C=C1