accessibility menu, dialog, popup

UserName

CAS RN 5578-73-4

ClCH3NOOOO

CAS RN 5578-73-4

IUPAC Name: 24-methyl-5,7,18,20-tetraoxa-24-azahexacyclo[11.11.0.0²,¹⁰.0⁴,⁸.0¹⁴,²².0¹⁷,²¹]tetracosa-1(13),2,4(8),9,11,14,16,21,23-nonaen-24-ium chloride
Synonyms: sanguinarine chloride
Molecular Weight (g/mol): 367.79
Molecular Formula: C20H14ClNO4
InChi Key: GIZKAXHWLRYMLE-UHFFFAOYSA-M
SMILES: [Cl-].C[N+]1=CC2=C3OCOC3=CC=C2C2=C1C1=CC3=C(OCO3)C=C1C=C2
Product Line 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results