accessibility menu, dialog, popup

UserName

ZnOOOO

CAS RN 557-41-5

IUPAC Name: zinc(2+) diformate
Synonyms: zinc(2+) diformate
Molecular Weight (g/mol): 155.41
Molecular Formula: C2H2O4Zn
InChi Key: XOBMCBQSUCOAOC-UHFFFAOYSA-L
SMILES: [Zn++].[O-]C=O.[O-]C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results