accessibility menu, dialog, popup

UserName

CAS RN 556-10-5

IUPAC Name: (4-nitrophenyl)urea
Synonyms: 4-nitrophenylurea
Molecular Weight (g/mol): 181.15
Molecular Formula: C7H7N3O3
InChi Key: LXXTVGKSGJADFU-UHFFFAOYSA-N
SMILES: NC(=O)NC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results