accessibility menu, dialog, popup

UserName

CAS RN 555-45-3

IUPAC Name: 1,3-bis(tetradecanoyloxy)propan-2-yl tetradecanoate
Synonyms: trimyristin
Molecular Weight (g/mol): 723.18
Molecular Formula: C45H86O6
InChi Key: DUXYWXYOBMKGIN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (1)
Quantity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)