missing translation for 'onlineSavingsMsg'
Learn More

Trimyristin, 95%

Catalog No. AAJ6281914
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6281914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6281914 Supplier Thermo Scientific Chemicals Supplier No. J6281914

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 555-45-3 | C45H86O6 | 723.177 g/mol

It can be used for the preparation of solid lipid nanoparticles (SLN) composed of trimyristin (solid lipid) and poloxamer 407 (surfactant). It is also used as a synthesis reagent and as an emollient, refatting agent, skin conditioning agent, solvents and viscosity controlling agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used for the preparation of solid lipid nanoparticles (SLN) composed of trimyristin (solid lipid) and poloxamer 407 (surfactant). It is also used as a synthesis reagent and as an emollient, refatting agent, skin conditioni

Chemical Identifiers

CAS 555-45-3
Molecular Formula C45H86O6
Molecular Weight (g/mol) 723.177
MDL Number MFCD00036229
InChI Key DUXYWXYOBMKGIN-UHFFFAOYSA-N
Synonym trimyristin, glycerol trimyristate, myristin, propane-1,2,3-triyl tritetradecanoate, glyceryl trimyristate, dynasan 114, myristic acid triglyceride, trimyristoylglycerol, tetradecanoic acid, 1,2,3-propanetriyl ester, myristin, tri
PubChem CID 11148
ChEBI CHEBI:77391
IUPAC Name 2,3-di(tetradecanoyloxy)propyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCC

Specifications

Melting Point 56°C to 57°C
Color White
Linear Formula [CH3(CH2)12COOCH2]2CHOCO(CH2)12CH3
Quantity 25 g
Beilstein 1718242
Merck Index 14,9724
Solubility Information Insoluble in water; Soluble in chloroform, ether, ethanol or dichloromethane
Formula Weight 723.16
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Trimyristin
Show More Show Less
EINECSNumber 209-099-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only