accessibility menu, dialog, popup

UserName

CH3H3CBOHHO

CAS RN 55499-43-9

IUPAC Name: (3,4-dimethylphenyl)boronic acid
Synonyms: 3,4-dimethylphenylboronic acid
Molecular Weight (g/mol): 149.98
Molecular Formula: C8H11BO2
InChi Key: KDVZJKOYSOFXRV-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1C)B(O)O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)