missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylphenylboronic acid, 97%, May contain varying amounts of anhydride, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals AC35928DA

Quantity 1g
Packaging Amber Glass Bottle

Catalog No. AC35928DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

55499-43-9
149.984
KDVZJKOYSOFXRV-UHFFFAOYSA-N
2734348
B(C1=CC(=C(C=C1)C)C)(O)O
C8H11BO2
MFCD01009694
3,4-dimethylphenyl boronic acid, 3,4-dimethylbenzeneboronic acid, 3,4-dimethylphenyl boranediol, 3,4-dimethylphenylboronicacid, 4-borono-o-xylene, boronic acid, 3,4-dimethylphenyl, pubchem1832, acmc-209lna, 3.4-dimethylbenzeneboronic acid, 3,4-dimethyl phenylboronic acid
(3,4-dimethylphenyl)boronic acid

Specifications

3, 4-Dimethylphenylboronic acid
96.0
Authentic
Amber Glass Bottle
1g
3,4-dimethylphenyl boronic acid, 3,4-dimethylbenzeneboronic acid, 3,4-dimethylphenyl boranediol, 3,4-dimethylphenylboronicacid, 4-borono-o-xylene, boronic acid, 3,4-dimethylphenyl, pubchem1832, acmc-209lna, 3.4-dimethylbenzeneboronic acid, 3,4-dimethyl phenylboronic acid
B(C1=CC(=C(C=C1)C)C)(O)O
149.984
149.98
55499-43-9
100.0
97%
C8H11BO2
MFCD01009694
KDVZJKOYSOFXRV-UHFFFAOYSA-N
(3,4-dimethylphenyl)boronic acid
2734348
≥96.0%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents