missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D31101G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D31101G Supplier TCI America Supplier No. D31101G

Specifications

Chemical Name or Material 3,4-Dimethylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 55499-43-9
Color White
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C8H11BO2
MDL Number MFCD01009694
Synonym 3,4-dimethylphenyl boronic acid, 3,4-dimethylbenzeneboronic acid, 3,4-dimethylphenyl boranediol, 3,4-dimethylphenylboronicacid, 4-borono-o-xylene, boronic acid, 3,4-dimethylphenyl, pubchem1832, acmc-209lna, 3.4-dimethylbenzeneboronic acid, 3,4-dimethyl phenylboronic acid
InChI Key KDVZJKOYSOFXRV-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1C)B(O)O
IUPAC Name (3,4-dimethylphenyl)boronic acid
Molecular Weight (g/mol) 149.98
PubChem CID 2734348
Formula Weight 149.98
Show More Show Less