accessibility menu, dialog, popup

UserName

CAS RN 55304-73-9

NOClCl

CAS RN 55304-73-9

IUPAC Name: 2,6-dichloropyridine-3-carbaldehyde
Synonyms: 2,6-dichloropyridine-3-carbaldehyde
Molecular Weight (g/mol): 176
Molecular Formula: C6H3Cl2NO
InChi Key: HWTMRGXKSANEDO-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=C(C=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results